Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tacc
Entrez Gene ID
40589
Gene Name
transforming acidic coiled-coil protein
Gene Aliases
CG9765, D-TACC, D-tacc, DTACC, Dmel\CG9765, Dmel_CG9765, TACC, TACCdm, acc, d-tacc, dTACC, dtacc
Chromosome Location
Chr.3R: 4733654 - 4749056 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0026620
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202238.2 NP_001189167.1 5 - 6 593 89
NM_001202239.2 NP_001189168.1 7 - 8 3306 89
NM_001202240.2 NP_001189169.1 7 - 8 3700 89
NM_001259986.1 NP_001246915.1 7 - 8 3227 89
NM_001275332.1 NP_001262261.1 7 - 8 3700 89
NM_001275333.1 NP_001262262.1 8 - 9 3541 89
NM_058085.5 NP_477433.2 7 - 8 3658 89
NM_058086.4 NP_477434.1 7 - 8 3345 89
NM_169016.3 NP_730867.2 7 - 8 3348 89
NM_176401.2 NP_788578.1 7 - 8 3613 89
NM_176402.3 NP_788579.1 5 - 6 551 89
NM_176403.2 NP_788580.1 5 - 6 544 89
NM_176404.2 NP_788581.1 5 - 6 586 89
GenBank mRNA AY058499.1 - 7 - 8 3658 89
BT099701.1 - 7 - 8 3314 89

View Full Product Details