Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rga
Entrez Gene ID
40683
Gene Name
Regena
Gene Aliases
CG2138, CG2161, Dmel\CG2161, Dmel_CG2161, NOT2, Not2, RGA, l(3)03834, not2, rga
Chromosome Location
Chr.3R: 5607275 - 5613112 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0017550
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300254.1 NP_001287183.1 6 - 7 2102 59
NM_079515.3 NP_524239.2 6 - 7 2404 59
NM_169080.3 NP_730966.1 6 - 7 2422 59
GenBank mRNA BT025208.2 - 6 - 7 2394 59

View Full Product Details