Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
alpha-Est10
Entrez Gene ID
40896
Gene Name
alpha-Esterase-10
Gene Aliases
CG1131, DmalphaE10, Dmel\CG1131, Dmel_CG1131, Esterase-10, K, aE10, alpha-Ests, alphaE10, anon-WO0172774.144
Chromosome Location
Chr.3R: 7486583 - 7493845 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015569
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260033.2 NP_001246962.1 2 - 3 435 101
NM_001260034.1 NP_001246963.1 2 - 3 381 101
NM_079533.4 NP_524257.3 2 - 3 581 101

View Full Product Details