Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- alpha-Est1
- Entrez Gene ID
-
40909
- Gene Name
- alpha-Esterase-1
- Gene Aliases
-
A, Alpha-Est1, CG1031, DmalphaE1, Dmel\CG1031, Dmel_CG1031, Est1, Q9VIC3_DROME, aE1, alpha est1, alpha-est1, alphaE1
- Chromosome Location
-
Chr.3R: 7545733 - 7549124 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0015568
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079545.4 |
NP_524269.3 |
2 - 3 |
699 |
85 |
GenBank mRNA |
AY058637.1 |
- |
2 - 3 |
678 |
85 |
View Full Product Details