Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Scgbeta
Entrez Gene ID
41525
Gene Name
Sarcoglycan beta
Gene Aliases
CG5657, DmSCG-beta, Dmel\CG5657, Dmel_CG5657, dscgbeta, scg-beta
Chromosome Location
Chr.3R: 12421236 - 12422429 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0038042
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079603.3 NP_524327.1 1 - 1 1138 58
GenBank mRNA AY071368.1 - 1 - 1 1138 58

View Full Product Details