Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
yellow-f
Entrez Gene ID
41595
Gene Name
CG18550-PA
Gene Aliases
CG18550, DI, Dmel\CG18550, Dmel_CG18550, Yellow-F, yell-f
Chromosome Location
Chr.3R: 12991449 - 12993201 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0041710
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275587.1 NP_001262516.1 3 - 4 1173 99
NM_001275588.1 NP_001262517.1 3 - 4 1158 99
NM_079611.2 NP_524335.1 3 - 4 1206 99

View Full Product Details