Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Crz
- Entrez Gene ID
-
41742
- Gene Name
- Corazonin
- Gene Aliases
-
CG3302, COR, CORZ, CRZ, Cora, Dm-Crz, Dmel\CG3302, Dmel_CG3302, Drm-COR, cor, crz
- Chromosome Location
-
Chr.3R: 14314854 - 14315716 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0013767
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079626.3 |
NP_524350.1 |
1 - 2 |
300 |
65 |
View Full Product Details