Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sap47
Entrez Gene ID
41938
Gene Name
Synapse-associated protein 47kD
Gene Aliases
CG8884, Dmel\CG8884, Dmel_CG8884, SAP 47, SAP-47, SAP47, dsap-47, sap47
Chromosome Location
Chr.3R: 15935100 - 15962124 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0013334
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275688.1 NP_001262617.1 2 - 3 855 74
NM_001300421.1 NP_001287350.1 1 - 2 1242 74
NM_001370053.1 NP_001356917.1 1 - 2 1242 74
NM_001370054.1 NP_001356918.1 2 - 3 855 74
NM_079647.4 NP_524371.1 1 - 2 1223 74
NM_169688.3 NP_732092.2 1 - 2 1242 74
NM_169689.3 NP_732093.1 2 - 3 855 74
GenBank mRNA BT126317.1 - 2 - 3 488 74
X80110.1 - - 514 74
X80111.1 - - 842 74

View Full Product Details