The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001260283.2 | NP_001247212.1 | 6 - 7 | 2713 | 78 |
NM_001260284.2 | NP_001247213.1 | 5 - 6 | 2568 | 78 | |
NM_001260285.2 | NP_001247214.1 | 5 - 6 | 2520 | 78 | |
NM_001260286.2 | NP_001247215.1 | 6 - 7 | 3431 | 78 | |
NM_001300505.1 | NP_001287434.1 | 5 - 6 | 2673 | 78 | |
NM_001300506.1 | NP_001287435.1 | 5 - 6 | 4087 | 78 | |
NM_079699.4 | NP_524423.2 | 4 - 5 | 3362 | 78 | |
NM_169940.4 | NP_732576.1 | 5 - 6 | 2765 | 78 | |
NM_169941.2 | NP_732577.1 | 4 - 5 | 2607 | 78 | |
GenBank mRNA | AY118680.1 | - | 5 - 6 | 2711 | 78 |
BT011420.1 | - | 5 - 6 | 2708 | 78 |