Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mvl
Entrez Gene ID
42490
Gene Name
Malvolio
Gene Aliases
CG3671, DMT1, Dmel\CG3671, Dmel_CG3671, NRM2, dNRAMP, mvl
Chromosome Location
Chr.3R: 21051383 - 21060800 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011672
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170197.1 NP_001163668.1 5 - 6 3037 65
NM_001260292.1 NP_001247221.1 6 - 7 921 65
NM_001275866.1 NP_001262795.1 6 - 7 908 65
NM_079701.5 NP_524425.2 6 - 7 1224 65
NM_169944.3 NP_732584.1 6 - 7 908 65
NM_206529.2 NP_996251.2 6 - 7 1224 65
GenBank mRNA AY058543.1 - 6 - 7 1183 65
BT044567.1 - 5 - 6 3035 65

View Full Product Details