Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pit
Entrez Gene ID
42595
Gene Name
pitchoune
Gene Aliases
CG6375, DmRH15, Dmel\CG6375, Dmel_CG6375, Hel93F, PIT, Pit, hel93F
Chromosome Location
Chr.3R: 22034298 - 22038925 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0266581
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300530.1 NP_001287459.1 4 - 5 1835 79
NM_079722.4 NP_524446.3 4 - 5 1835 79
NM_169991.2 NP_732694.2 5 - 6 1889 79
GenBank mRNA AY071402.1 - 5 - 6 1889 79
AY119620.1 - 4 - 5 1811 79

View Full Product Details