Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EndoA
Entrez Gene ID
42265
Gene Name
Endophilin A
Gene Aliases
CG14296, D-endo, D-endoA, Dendo-A, Dmel\CG14296, Dmel_CG14296, Endo, endo, endoA
Chromosome Location
Chr.3R: 18906609 - 18910755 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0038659
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275788.1 NP_001262717.1 3 - 3 3303 77
NM_138767.3 NP_620122.2 1 - 1 3537 77
NM_169838.2 NP_732383.1 2 - 2 3358 77
GenBank mRNA BT044220.1 - 1 - 1 3531 77

View Full Product Details