Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Snm1
Entrez Gene ID
40669
Gene Name
CG10018-PA
Gene Aliases
CG10018, Dmel\CG10018, Dmel_CG10018, SNM1, mus322
Chromosome Location
Chr.3R: 5555084 - 5557786 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037338
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_141291.4 NP_649548.1 1 - 2 1113 73
GenBank mRNA AY058585.1 - 1 - 2 1085 73

View Full Product Details