The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_141298.2 | NP_649555.1 | 1 - 1 | 596 | 78 |
GenBank mRNA | AB115908.1 | - | 1 - 1 | 526 | 78 |