Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Osi3
Entrez Gene ID
40757
Gene Name
Osiris 3
Gene Aliases
CG1150, Dmel\CG1150, Dmel_CG1150, osi3
Chromosome Location
Chr.3R: 6215832 - 6218199 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037411
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275375.1 NP_001262304.1 1 - 2 315 79
NM_141365.3 NP_649622.2 1 - 2 315 79
GenBank mRNA BT044145.1 - 1 - 2 311 79

View Full Product Details