Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Osi13
Entrez Gene ID
40769
Gene Name
Osiris 13
Gene Aliases
CG15595, Dmel\CG15595, Dmel_CG15595
Chromosome Location
Chr.3R: 6291737 - 6293489 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037422
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_141377.2 NP_649634.1 1 - 2 438 95
GenBank mRNA BT122093.2 - 1 - 2 273 95

View Full Product Details