Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Neurochondrin
Entrez Gene ID
40796
Gene Name
CG2330-PA
Gene Aliases
CG2330, Dmel\CG2330, Dmel_CG2330
Chromosome Location
Chr.3R: 6468437 - 6471016 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037447
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_141401.2 NP_649658.1 1 - 1 2414 92

View Full Product Details