Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glob2
Entrez Gene ID
40807
Gene Name
globin 2
Gene Aliases
CG15180, DmeGb2, Dmel\CG15180, Dmel_CG15180, anon-SAGE:Wang-121
Chromosome Location
Chr.3R: 6605997 - 6608914 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0250846
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275393.1 NP_001262322.1 1 - 2 325 65

View Full Product Details