Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glob2
Entrez Gene ID
40807
Gene Name
globin 2
Gene Aliases
CG15180, DmeGb2, Dmel\CG15180, Dmel_CG15180, anon-SAGE:Wang-121
Chromosome Location
Chr.3R: 6605997 - 6608914 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0250846
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275392.2 NP_001262321.1 2 - 3 524 82
NM_001275393.1 NP_001262322.1 2 - 3 485 82
NM_141412.3 NP_649669.3 2 - 3 524 82
GenBank mRNA AM086024.1 - 2 - 3 448 82
BT022295.1 - 2 - 3 469 82
BT133287.1 - 2 - 3 543 82

View Full Product Details