Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DNApol-iota
Entrez Gene ID
40965
Gene Name
DNA polymerase iota
Gene Aliases
AAF54198, CG7602, DNA Pol II-iota, DNA-pol iota, DNApol-i, Dm-Poliota, Dmel\CG7602, Dmel_CG7602, Poliota, drad30B
Chromosome Location
Chr.3R: 8233507 - 8237389 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037554
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300298.1 NP_001287227.1 2 - 3 1045 77
NM_141515.3 NP_649772.1 2 - 3 1110 77
NM_169207.2 NP_731214.1 2 - 3 1032 77
GenBank mRNA AB049434.1 - 2 - 3 1039 77
AY051762.1 - 2 - 3 1043 77

View Full Product Details