Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RagA-B
- Entrez Gene ID
-
41071
- Gene Name
- Ras-related GTP binding A/B
- Gene Aliases
-
CG11968, Dmel\CG11968, Dmel_CG11968, Gtr1, Rag, Rag A, RagA, RagA/B, dRagA, dRagB, rag, ragA
- Chromosome Location
-
Chr.3R: 8982732 - 8984315 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0037647
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_141593.4 |
NP_649850.1 |
3 - 4 |
590 |
70 |
GenBank mRNA |
AY047538.1 |
- |
3 - 4 |
569 |
70 |
View Full Product Details