Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pic
Entrez Gene ID
41611
Gene Name
piccolo
Gene Aliases
0263/16, 1258/16, 1366/03, 153636_at, 263/16, CG7769, D-DDB1, DDB-1, DDB1, Dbd1, Ddb1, Dmel\CG7769, Dmel_CG7769, PIC, d-ddb1, dDDB1, dDdb1, ddb1, l(3)87Dh, l(3)S136603
Chromosome Location
Chr.3R: 13052245 - 13056725 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0260962
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_142000.3 NP_650257.1 3 - 4 1556 69
GenBank mRNA AF132145.1 - 3 - 4 1538 69

View Full Product Details