Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- wntD
- Entrez Gene ID
-
41633
- Gene Name
- wnt inhibitor of Dorsal
- Gene Aliases
-
CG8458, DWnt-8, DWnt8, Dm DWnt8, Dmel\CG8458, Dmel_CG8458, Dwnt-8, Dwnt-D, Dwnt8, Wnt8, WntD, wnt8
- Chromosome Location
-
Chr.3R: 13292052 - 13293198 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0038134
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_142015.2 |
NP_650272.1 |
1 - 1 |
1044 |
149 |
GenBank mRNA |
AY071538.1 |
- |
1 - 1 |
1044 |
149 |
View Full Product Details