Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PKD
Entrez Gene ID
42203
Gene Name
Protein Kinase D
Gene Aliases
150131_at, CG7125, Dmel\CG7125, Dmel_CG7125, PKCm, dPKD
Chromosome Location
Chr.3R: 18303279 - 18314407 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0038603
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144602.2 NP_001138074.1 7 - 8 2827 78
NM_001275775.1 NP_001262704.1 7 - 8 2924 78
NM_001275776.1 NP_001262705.1 7 - 8 2850 78
NM_142453.4 NP_650710.3 7 - 8 2960 78
NM_169803.4 NP_732325.3 7 - 8 2850 78
NM_169804.4 NP_732326.3 7 - 8 2859 78
NM_169805.4 NP_732327.3 7 - 8 2878 78
GenBank mRNA AY118312.1 - 7 - 8 2960 78

View Full Product Details