Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Nup58
- Entrez Gene ID
-
42342
- Gene Name
- Nucleoporin 58kD
- Gene Aliases
-
CG7360, Dm NUP58, Dmel\CG7360, Dmel_CG7360, RE52572p, nup58
- Chromosome Location
-
Chr.3R: 19645026 - 19646906 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0038722
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_142564.4 |
NP_650821.2 |
1 - 2 |
1059 |
68 |
View Full Product Details