Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Rassf
- Entrez Gene ID
-
42734
- Gene Name
- Ras association family member
- Gene Aliases
-
CG4656, Dmel\CG4656, Dmel_CG4656, RASSF, dRASSF, dRASSF1, dRassf, dmRASSF, rassf
- Chromosome Location
-
Chr.3R: 23114014 - 23117931 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0039055
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_142869.5 |
NP_651126.3 |
7 - 8 |
2759 |
58 |
GenBank mRNA |
AY051923.1 |
- |
7 - 8 |
2724 |
58 |
View Full Product Details