Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Exo84
Entrez Gene ID
43163
Gene Name
Exocyst 84
Gene Aliases
CG6095, DExo84, Dmel\CG6095, Dmel_CG6095, Exo84p, anon-WO0153538.55, exo84, onr
Chromosome Location
Chr.3R: 26049731 - 26052257 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0266668
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_143197.2 NP_651454.1 3 - 4 555 66
NM_206576.2 NP_996299.1 3 - 4 558 66

View Full Product Details