Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TwdlO
Entrez Gene ID
43204
Gene Name
TweedleO
Gene Aliases
CG6452, Dmel\CG6452, Dmel_CG6452
Chromosome Location
Chr.3R: 26625932 - 26626738 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039438
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_143230.2 NP_651487.1 1 - 1 160 65
GenBank mRNA BT100067.4 - 1 - 1 119 65

View Full Product Details