Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- mfrn
- Entrez Gene ID
-
43353
- Gene Name
- mitoferrin
- Gene Aliases
-
CG4963, Dmel\CG4963, Dmel_CG4963, HD35059, Mfrn, anon-EST:fe2H5, dmfrn
- Chromosome Location
-
Chr.3R: 27942796 - 27948021 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0039561
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_143343.3 |
NP_651600.1 |
2 - 3 |
921 |
98 |
GenBank mRNA |
AY060268.1 |
- |
2 - 3 |
901 |
98 |
View Full Product Details