Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
yata
Entrez Gene ID
43508
Gene Name
CG1973-PA
Gene Aliases
CG1973, Dmel\CG1973, Dmel_CG1973, Yata
Chromosome Location
Chr.3R: 29749219 - 29754528 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0260990
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276153.1 NP_001263082.1 6 - 7 2534 95
NM_143465.2 NP_651722.1 6 - 7 2534 95
GenBank mRNA BT032863.1 - 6 - 7 2518 95

View Full Product Details