Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Osi23
- Entrez Gene ID
-
43615
- Gene Name
- Osiris 23
- Gene Aliases
-
CG15538, Dmel\CG15538, Dmel_CG15538, gox
- Chromosome Location
-
Chr.3R: 30444076 - 30445046 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0039771
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_143537.3 |
NP_651794.2 |
2 - 3 |
629 |
97 |
View Full Product Details