Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Smvt
- Entrez Gene ID
-
43744
- Gene Name
- Sodium-dependent multivitamin transporter
- Gene Aliases
-
CG2191, Dmel\CG2191, Dmel_CG2191, SMVT, dSLC5A5, smvt
- Chromosome Location
-
Chr.3R: 31759404 - 31761579 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0039873
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_143634.4 |
NP_651891.2 |
1 - 2 |
493 |
70 |
GenBank mRNA |
BT050561.1 |
- |
1 - 2 |
493 |
70 |
View Full Product Details