Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MED6
Entrez Gene ID
44728
Gene Name
Mediator complex subunit 6
Gene Aliases
BcDNA:LD15729, CG9473, Dmel\CG9473, Dmel_CG9473, Med6, Trap32, anon-EST:Posey279, dMED6, dMed6, dTRAP32, med6, p35
Chromosome Location
Chr.3R: 9766031 - 9768984 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0024330
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170106.1 NP_001163577.1 2 - 3 796 80
NM_001170107.1 NP_001163578.1 2 - 3 840 80
NM_001300335.1 NP_001287264.1 1 - 2 643 80
NM_143731.6 NP_651988.2 2 - 3 845 80
NM_169310.2 NP_731403.2 2 - 3 784 80
GenBank mRNA AY118517.1 - 2 - 3 822 80
BT001633.1 - 2 - 3 784 80

View Full Product Details