Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PpD3
Entrez Gene ID
49779
Gene Name
Protein phosphatase D3
Gene Aliases
CG8402, D3, DmPP5, DmPpp5-85E, Dmel\CG8402, Dmel_CG8402, PP5, PP5 85E, Pp5-85E
Chromosome Location
Chr.3R: 9747875 - 9751094 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0005777
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275504.2 NP_001262433.1 6 - 7 1507 129
NM_080207.3 NP_524946.1 7 - 8 1612 129
NM_169307.2 NP_731398.1 7 - 8 1477 129
GenBank mRNA AJ271781.1 - 6 - 7 1426 129
AY058356.1 - 6 - 7 1470 129

View Full Product Details