Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Men
Entrez Gene ID
47173
Gene Name
Malic enzyme
Gene Aliases
CG10120, Dmel\CG10120, Dmel_CG10120, MDH, ME, MEN, Mdh-NADP, Me, anon-WO0118547.278, mem, men
Chromosome Location
Chr.3R: 12713103 - 12722442 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0002719
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275580.1 NP_001262509.1 2 - 3 1319 53
NM_080141.4 NP_524880.2 3 - 4 1861 53
NM_169479.2 NP_731739.1 3 - 4 1722 53
GenBank mRNA AY118540.1 - 3 - 4 1964 53
BT100054.1 - 3 - 4 1709 53
DQ148978.1 - 2 - 3 1061 53
DQ148979.1 - 2 - 3 1061 53
DQ148980.1 - 2 - 3 1061 53
DQ148981.1 - 2 - 3 1061 53
DQ148982.1 - 2 - 3 1061 53
DQ148983.1 - 2 - 3 1061 53
DQ148988.1 - 2 - 3 1061 53
DQ148991.1 - 2 - 3 1061 53
DQ148992.1 - 2 - 3 1061 53
DQ148994.1 - 2 - 3 1061 53
DQ148997.1 - 2 - 3 1061 53
DQ148998.1 - 2 - 3 1061 53
DQ149000.1 - 2 - 3 1061 53
DQ149001.1 - 2 - 3 1061 53
DQ149002.1 - 2 - 3 1061 53
DQ149003.1 - 2 - 3 1061 53
DQ149004.1 - 2 - 3 1061 53
DQ149005.1 - 2 - 3 1061 53
DQ149006.1 - 2 - 3 1061 53
DQ149007.1 - 2 - 3 1061 53
DQ149008.1 - 2 - 3 1061 53
DQ149009.1 - 2 - 3 1061 53
JQ690827.1 - 2 - 3 1097 53
JQ690831.1 - 2 - 3 1097 53
JQ690840.1 - 2 - 3 1073 53
JQ690841.1 - 2 - 3 1097 53
JQ690842.1 - 2 - 3 1097 53
JQ690843.1 - 2 - 3 1097 53
JQ690854.1 - 2 - 3 1097 53
JQ690856.1 - 2 - 3 1097 53
JQ690859.1 - 2 - 3 1097 53

View Full Product Details