Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sap47
Entrez Gene ID
41938
Gene Name
Synapse-associated protein 47kD
Gene Aliases
CG8884, Dmel\CG8884, Dmel_CG8884, SAP 47, SAP-47, SAP47, dsap-47, sap47
Chromosome Location
Chr.3R: 15935100 - 15962124 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0013334
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_169684.3 NP_732088.1 1 - 2 1219 65
NM_169685.3 NP_732089.1 2 - 3 827 65
NM_169686.3 NP_732090.1 2 - 3 1155 65
NM_169687.3 NP_732091.1 2 - 3 973 65
NM_169690.3 NP_732094.1 1 - 2 1219 65
GenBank mRNA AY051871.1 - 1 - 2 1167 65

View Full Product Details