Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ir94a
- Entrez Gene ID
-
318610
- Gene Name
- Ionotropic receptor 94a
- Gene Aliases
-
CG31164, DmelIR94a, Dmel\CG31164, Dmel_CG31164, IR94a
- Chromosome Location
-
Chr.3R: 22113427 - 22115214 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0051164
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_169993.1 |
NP_732699.1 |
1 - 1 |
1719 |
91 |
View Full Product Details