Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GluProRS
- Entrez Gene ID
-
42834
- Gene Name
- Glutamyl-prolyl-tRNA synthetase
- Gene Aliases
-
AATS, AATs-GluPro, Aa-tRNA-syn-glupro, Aats-glu, Aats-glupro, CG5394, Dmel\CG5394, Dmel_CG5394, EPRS, ERS, FBpp0083898, Glu-ProRS, GluPro-RS, GluRS, glutamyl-prolyl-tRNA synthetase, p200
- Chromosome Location
-
Chr.3R: 23952136 - 23958391 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0005674
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_170103.2 |
NP_732925.1 |
1 - 2 |
44 |
65 |
View Full Product Details