Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ppk22
- Entrez Gene ID
-
318593
- Gene Name
- pickpocket 22
- Gene Aliases
-
CG31105, Dmel\CG31105, Dmel_CG31105, FBgn0051105
- Chromosome Location
-
Chr.3R: 24857585 - 24859801 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0051105
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_170175.4 |
NP_733051.2 |
7 - 8 |
1596 |
68 |
View Full Product Details