Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHKov1
Entrez Gene ID
43067
Gene Name
CHK domain ov1
Gene Aliases
BcDNA:GH03753, CG10618, Dmel\CG10618, Dmel_CG10618, ref(3)D
Chromosome Location
Chr.3R: 25323156 - 25329316 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0045761
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104448.2 NP_001097918.1 3 - 4 755 91

View Full Product Details