Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ser
Entrez Gene ID
43275
Gene Name
Serrate
Gene Aliases
Bd, CG6127, CT18858, Dmel\CG6127, Dmel_CG6127, Rpw, SER, mel(3)8, ser, std
Chromosome Location
Chr.3R: 27172090 - 27194267 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004197
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300629.1 NP_001287558.1 11 - 12 3096 74
NM_079803.3 NP_524527.3 11 - 12 2824 74

View Full Product Details