Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ser
Entrez Gene ID
43275
Gene Name
Serrate
Gene Aliases
Bd, CG6127, CT18858, Dmel\CG6127, Dmel_CG6127, Rpw, SER, mel(3)8, ser, std
Chromosome Location
Chr.3R: 27172090 - 27194267 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004197
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300629.1 NP_001287558.1 6 - 7 1621 71
NM_079803.3 NP_524527.3 6 - 7 1349 71

View Full Product Details