Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
side
Entrez Gene ID
43300
Gene Name
sidestep
Gene Aliases
CG12510, CG13973, CG13974, CG13975, CG13979, CG31062, CT33527, CT33528, Dmel\CG31062, Dmel_CG31062, Doe, Side, doe
Chromosome Location
Chr.3R: 27375088 - 27437482 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0016061
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144645.3 NP_001138117.1 7 - 8 2446 66
NM_001260405.2 NP_001247334.1 7 - 8 2135 66
NM_001260406.2 NP_001247335.1 7 - 8 2868 66
NM_079806.4 NP_524530.2 7 - 8 2868 66

View Full Product Details