Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
side
Entrez Gene ID
43300
Gene Name
sidestep
Gene Aliases
CG12510, CG13973, CG13974, CG13975, CG13979, CG31062, CT33527, CT33528, Dmel\CG31062, Dmel_CG31062, Doe, Side, doe
Chromosome Location
Chr.3R: 27375088 - 27437482 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0016061
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144645.3 NP_001138117.1 10 - 11 2854 93
NM_001260405.2 NP_001247334.1 10 - 11 2543 93
NM_001260406.2 NP_001247335.1 10 - 11 3276 93
NM_079806.4 NP_524530.2 10 - 11 3276 93

View Full Product Details