The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170296.2 | NP_001163767.1 | 3 - 4 | 735 | 103 |
NM_001300647.1 | NP_001287576.1 | 3 - 4 | 735 | 103 | |
NM_001300648.1 | NP_001287577.1 | 3 - 4 | 769 | 103 | |
NM_079821.4 | NP_524545.2 | 3 - 4 | 735 | 103 | |
GenBank mRNA | BT023513.1 | - | 3 - 4 | 704 | 103 |