Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PH4alphaMP
- Entrez Gene ID
-
43624
- Gene Name
- prolyl-4-hydroxylase-alpha MP
- Gene Aliases
-
59D7T, CG9726, DM59D7T, Dmel\CG9726, Dmel_CG9726, ESTS:59D7T
- Chromosome Location
-
Chr.3R: 30491813 - 30496153 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0026190
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079855.3 |
NP_524594.2 |
5 - 6 |
1575 |
74 |
GenBank mRNA |
BT132909.1 |
- |
5 - 6 |
1574 |
74 |
View Full Product Details