Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- mRpL32
- Entrez Gene ID
-
43698
- Gene Name
- mitochondrial ribosomal protein L32
- Gene Aliases
-
BcDNA:LD41522, CG12220, Dmel\CG12220, Dmel_CG12220, L32
- Chromosome Location
-
Chr.3R: 31252872 - 31253686 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0039835
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079867.3 |
NP_524606.1 |
1 - 2 |
130 |
69 |
GenBank mRNA |
AY118962.1 |
- |
1 - 2 |
100 |
69 |
View Full Product Details