Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SF1
Entrez Gene ID
43912
Gene Name
Splicing factor 1
Gene Aliases
CG5836, Dmel\CG5836, Dmel_CG5836, cg5836, dSF1, p70
Chromosome Location
Chr.3R: 17540421 - 17545148 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0025571
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275743.2 NP_001262672.1 2 - 3 992 74
NM_001275744.1 NP_001262673.1 2 - 3 992 74
NM_079915.3 NP_524654.2 2 - 3 992 74
GenBank mRNA BT031265.1 - 2 - 3 903 74

View Full Product Details