Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- woc
- Entrez Gene ID
-
47249
- Gene Name
- without children
- Gene Aliases
-
CG5965, Dmel\CG5965, Dmel_CG5965, WOC, Woc
- Chromosome Location
-
Chr.3R: 27256595 - 27264055 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0010328
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080147.3 |
NP_524886.2 |
3 - 4 |
1475 |
69 |
View Full Product Details