Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- wts
- Entrez Gene ID
-
43651
- Gene Name
- warts
- Gene Aliases
-
CG12072, Dlats, Dmel\CG12072, Dmel_CG12072, LATS, LATS1, Lats, Lats/Warts, MENE (3R)-G, WTS, WTS/LATS, Warts, Warts/Lats, Wts, Wts/Lats, dmLATS, l(3)100Aa, lats, lts, wart, warts/lats, wts/lats
- Chromosome Location
-
Chr.3R: 30789625 - 30806619 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0011739
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_170524.2 |
NP_733403.1 |
4 - 5 |
3662 |
66 |
View Full Product Details